Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005632.2 | NP_005623.1 | 9, 10 | 2705 | ||
XM_011522620.2 | XP_011520922.1 | 10, 11 | 6004 | |||
XM_011522621.2 | XP_011520923.1 | 10, 11 | 6932 | |||
XM_011522622.1 | XP_011520924.1 | 8, 9 | 5636 | |||
XM_011522623.2 | XP_011520925.1 | 11, 12 | 3707 | |||
XM_011522624.2 | XP_011520926.1 | 10, 11 | 3045 | |||
XM_011522625.2 | XP_011520927.1 | 9, 10 | 2931 | |||
XM_011522626.2 | XP_011520928.1 | 10, 11 | 6496 | |||
XM_011522627.2 | XP_011520929.1 | 10, 11 | 6501 | |||
XM_011522628.2 | XP_011520930.1 | 10, 11 | 3565 | |||
XM_011522629.2 | XP_011520931.1 | 10, 11 | 3428 | |||
XM_011522630.2 | XP_011520932.1 | 9, 10 | 2766 | |||
XM_011522631.2 | XP_011520933.1 | 8, 9 | 2652 | |||
XM_011522632.2 | XP_011520934.1 | 8, 9 | 3083 | |||
XM_017023596.1 | XP_016879085.1 | 11, 12 | 3844 | |||
GenBank mRNA | BC021854.1 | - | 5, 6 | 772 | ||
U85647.1 | - | 9, 10 | 2705 |