Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030288.2 | NP_001025459.1 | 2 | 530 | ||
NM_003123.4 | NP_003114.1 | 2 | 497 | |||
GenBank mRNA | AK292626.1 | - | 2 | 469 | ||
AK313750.1 | - | 2 | 469 | |||
BC012350.1 | - | 2 | 417 | 4564347 | ||
DA951208.1 | - | 2 | 530 | |||
DB108930.1 | - | 2 | 497 | |||
J04536.1 | - | 2 | 457 | |||
X60502.1 | - | 2 | 501 |