Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003136.3 | NP_003127.1 | 3 | 471 | ||
XM_005268024.2 | XP_005268081.1 | 3 | 724 | |||
XM_011537106.1 | XP_011535408.1 | 3 | 352 | |||
XM_017021615.1 | XP_016877104.1 | 3 | 396 | |||
GenBank mRNA | AK312853.1 | - | 3 | 416 | ||
BC000652.2 | - | 3 | 338 | 3343314 | ||
BC003389.1 | - | 3 | 344 | 3446599 | ||
U51920.1 | - | 3 | 191 |