Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003155.2 | NP_003146.1 | 2 | 500 | ||
GenBank mRNA | AK297734.1 | - | 3 | 347 | ||
AK309366.1 | - | 2 | 364 | |||
AK312589.1 | - | 2 | 499 | |||
BC029044.1 | - | 2 | 478 | 5191420 | ||
KP455655.1 | - | 2 | 163 | |||
KP455656.1 | - | 2 | 163 | |||
KP455657.1 | - | 2 | 163 | |||
KP455658.1 | - | 2 | 164 | |||
KP455659.1 | - | 2 | 163 | |||
U25997.1 | - | 2 | 500 | |||
U46768.1 | - | 2 | 250 |