Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001058.3 | NP_001049.1 | 2 | 1122 | ||
NM_015727.2 | NP_056542.1 | 2 | 1122 | |||
GenBank mRNA | AK289765.1 | - | 2 | 1091 | ||
AY462098.1 | - | 2 | 537 | |||
BC074911.2 | - | 2 | 559 | 30915523 | ||
BC074912.2 | - | 2 | 559 | 30915310 | ||
BC095528.1 | - | 2 | 534 | |||
M74290.1 | - | 2 | 747 | |||
M76675.1 | - | 2 | 537 | |||
M81797.1 | - | 2 | 581 | |||
M84425.1 | - | 2 | 659 | |||
M84426.1 | - | 2 | 659 | |||
S62045.1 | - | 2 | 601 |
Set Membership: |
![]() |