Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
siRNA ID
HSS110533
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
12, 13
1595
NM_001243226.2
NP_001230155.2
13, 14
1385
NM_001243227.1
NP_001230156.1
11, 12
1429
NM_001243228.1
NP_001230157.1
12, 13
1539
NM_001243230.1
NP_001230159.1
11, 12
1071
NM_001243231.1
NP_001230160.1
10, 11
1029
NM_001243232.1
NP_001230161.1
8, 9
1051
NM_001243233.1
NP_001230162.1
8, 9
992
NM_001243234.1
NP_001230163.1
5, 6
624
NM_001243235.1
NP_001230164.1
5, 6
624
NM_001243236.1
NP_001230165.1
5, 6
640
NM_001306207.1
NP_001293136.1
11, 12
1220
NM_001306208.1
NP_001293137.1
8, 9
1051
NM_003199.2
NP_003190.1
12, 13
1595
XM_005266739.3
XP_005266796.2
11, 12
1235
XM_005266741.3
XP_005266798.1
12, 13
1195
XM_005266743.4
XP_005266800.1
11, 12
1056
XM_005266744.3
XP_005266801.1
11, 12
1176
XM_005266745.3
XP_005266802.1
11, 12
1038
XM_005266747.3
XP_005266804.1
10, 11
1032
XM_005266749.3
XP_005266806.1
8, 9
823
XM_005266752.4
XP_005266809.1
8, 9
1393
XM_005266754.3
XP_005266811.1
7, 8
817
XM_005266755.4
XP_005266812.1
8, 9
852
XM_005266761.3
XP_005266818.1
5, 6
1136
XM_006722536.2
XP_006722599.1
12, 13
1518
XM_006722537.2
XP_006722600.1
12, 13
1151
XM_006722538.2
XP_006722601.1
11, 12
1173
XM_011526158.2
XP_011524460.1
11, 12
2149
XM_011526160.2
XP_011524462.1
7, 8
1202
XM_017025934.1
XP_016881423.1
11, 12
1232
XM_017025935.1
XP_016881424.1
11, 12
1235
XM_017025936.1
XP_016881425.1
11, 12
1235
XM_017025937.1
XP_016881426.1
11, 12
989
XM_017025938.1
XP_016881427.1
12, 13
1148
XM_017025939.1
XP_016881428.1
12, 13
1198
XM_017025940.1
XP_016881429.1
12, 13
1518
XM_017025941.1
XP_016881430.1
12, 13
1151
XM_017025942.1
XP_016881431.1
12, 13
1195
XM_017025943.1
XP_016881432.1
12, 13
1198
XM_017025944.1
XP_016881433.1
11, 12
1453
XM_017025945.1
XP_016881434.1
11, 12
1456
XM_017025946.1
XP_016881435.1
11, 12
1470
XM_017025947.1
XP_016881436.1
11, 12
1056
XM_017025948.1
XP_016881437.1
11, 12
1453
XM_017025949.1
XP_016881438.1
11, 12
1173
XM_017025950.1
XP_016881439.1
11, 12
1453
XM_017025951.1
XP_016881440.1
10, 11
1032
XM_017025952.1
XP_016881441.1
8, 9
1046
XM_017025953.1
XP_016881442.1
8, 9
823
XM_017025954.1
XP_016881443.1
8, 9
1407
XM_017025955.1
XP_016881444.1
7, 8
1141
XM_017025956.1
XP_016881445.1
7, 8
817
XM_017025957.1
XP_016881446.1
5, 6
1136
GenBank mRNA
AB209741.1
-
7, 8
703
AK095041.1
-
8, 9
1051
AK096862.1
-
10, 11
1029
AK122765.1
-
12, 13
1595
AK299169.1
-
11, 12
1429
AK300038.1
-
5, 6
570
AK300612.1
-
8, 9
992
AK300636.1
-
7, 8
722
AK301144.1
-
11, 12
1206
AK315074.1
-
12, 13
1527
AK316165.1
-
7, 8
766
AL518903.3
-
5, 6
640
BC031056.1
-
13, 14
1346
BC125084.1
-
11, 12
1009
40117795
BC125085.1
-
11, 12
1009
40117798
FR748210.1
-
11, 12
983
FR748211.1
-
11, 12
983
FR748212.1
-
9, 10
803
FR748213.1
-
9, 10
803
FR748214.1
-
10, 11
911
FR748215.1
-
8, 9
731
FR748216.1
-
11, 12
977
FR748217.1
-
10, 11
857
FR748218.1
-
6, 7
593
FR748219.1
-
8, 9
770
FR748220.1
-
5, 6
503
FR748221.1
-
5, 6
503
FR748222.1
-
5, 6
503
FR748223.1
-
3, 4
335
M74718.1
-
5, 6
455
M74719.1
-
12, 13
1182
X52079.1
-
9, 10
848
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0