Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
siRNA ID
HSS110535
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
11
1421
NM_001243226.2
NP_001230155.2
12
1211
NM_001243227.1
NP_001230156.1
10
1255
NM_001243228.1
NP_001230157.1
11
1365
NM_001243230.1
NP_001230159.1
10
897
NM_001243231.1
NP_001230160.1
9
855
NM_001243232.1
NP_001230161.1
7
877
NM_001243233.1
NP_001230162.1
7
818
NM_001243234.1
NP_001230163.1
4
450
NM_001243235.1
NP_001230164.1
4
450
NM_001243236.1
NP_001230165.1
4
466
NM_001306207.1
NP_001293136.1
10
1046
NM_001306208.1
NP_001293137.1
7
877
NM_003199.2
NP_003190.1
11
1421
XM_005266739.3
XP_005266796.2
10
1061
XM_005266741.3
XP_005266798.1
11
1021
XM_005266743.4
XP_005266800.1
10
882
XM_005266744.3
XP_005266801.1
10
1002
XM_005266745.3
XP_005266802.1
10
864
XM_005266747.3
XP_005266804.1
9
858
XM_005266749.3
XP_005266806.1
7
649
XM_005266752.4
XP_005266809.1
7
1219
XM_005266754.3
XP_005266811.1
6
643
XM_005266755.4
XP_005266812.1
7
678
XM_005266761.3
XP_005266818.1
4
962
XM_006722536.2
XP_006722599.1
11
1344
XM_006722537.2
XP_006722600.1
11
977
XM_006722538.2
XP_006722601.1
10
999
XM_011526158.2
XP_011524460.1
10
1975
XM_011526160.2
XP_011524462.1
6
1028
XM_017025934.1
XP_016881423.1
10
1058
XM_017025935.1
XP_016881424.1
10
1061
XM_017025936.1
XP_016881425.1
10
1061
XM_017025937.1
XP_016881426.1
10
815
XM_017025938.1
XP_016881427.1
11
974
XM_017025939.1
XP_016881428.1
11
1024
XM_017025940.1
XP_016881429.1
11
1344
XM_017025941.1
XP_016881430.1
11
977
XM_017025942.1
XP_016881431.1
11
1021
XM_017025943.1
XP_016881432.1
11
1024
XM_017025944.1
XP_016881433.1
10
1279
XM_017025945.1
XP_016881434.1
10
1282
XM_017025946.1
XP_016881435.1
10
1296
XM_017025947.1
XP_016881436.1
10
882
XM_017025948.1
XP_016881437.1
10
1279
XM_017025949.1
XP_016881438.1
10
999
XM_017025950.1
XP_016881439.1
10
1279
XM_017025951.1
XP_016881440.1
9
858
XM_017025952.1
XP_016881441.1
7
872
XM_017025953.1
XP_016881442.1
7
649
XM_017025954.1
XP_016881443.1
7
1233
XM_017025955.1
XP_016881444.1
6
967
XM_017025956.1
XP_016881445.1
6
643
XM_017025957.1
XP_016881446.1
4
962
GenBank mRNA
AB209741.1
-
6
529
AK095041.1
-
7
877
AK096862.1
-
9
855
AK122765.1
-
11
1421
AK299169.1
-
10
1255
AK300038.1
-
4
396
AK300612.1
-
7
818
AK300636.1
-
6
548
AK301144.1
-
10
1032
AK315074.1
-
11
1353
AK316165.1
-
6
592
AL518903.3
-
4
466
BC031056.1
-
12
1172
BC125084.1
-
10
835
40117795
BC125085.1
-
10
835
40117798
DC298029.1
-
4
450
FR748210.1
-
10
809
FR748211.1
-
10
809
FR748212.1
-
8
629
FR748213.1
-
8
629
FR748214.1
-
9
737
FR748215.1
-
7
557
FR748216.1
-
10
803
FR748217.1
-
9
683
FR748218.1
-
5
419
FR748219.1
-
7
596
FR748220.1
-
4
329
FR748221.1
-
4
329
FR748222.1
-
4
329
FR748223.1
-
2
161
M74718.1
-
4
281
M74719.1
-
11
1008
X52079.1
-
8
674
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-tm9kw:80/10.239.162.147:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0