Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
siRNA ID
HSS110549
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
2808
NM_001174093.1
NP_001167564.1
6
2468
NM_001174094.1
NP_001167565.1
7
2805
NM_001174095.1
NP_001167566.1
6
2327
NM_001174096.1
NP_001167567.1
7
2531
NM_001323638.1
NP_001310567.1
11
3178
NM_001323641.1
NP_001310570.1
7
2758
NM_001323642.1
NP_001310571.1
9
3060
NM_001323643.1
NP_001310572.1
9
2838
NM_001323644.1
NP_001310573.1
6
2557
NM_001323645.1
NP_001310574.1
8
2901
NM_001323646.1
NP_001310575.1
10
3139
NM_001323647.1
NP_001310576.1
8
2671
NM_001323648.1
NP_001310577.1
7
2611
NM_001323649.1
NP_001310578.1
8
2674
NM_001323650.1
NP_001310579.1
10
2906
NM_001323651.1
NP_001310580.1
8
2574
NM_001323652.1
NP_001310581.1
9
2723
NM_001323653.1
NP_001310582.1
9
2830
NM_001323654.1
NP_001310583.1
9
2717
NM_001323655.1
NP_001310584.1
9
2827
NM_001323656.1
NP_001310585.1
7
2502
NM_001323657.1
NP_001310586.1
7
2505
NM_001323658.1
NP_001310587.1
7
2344
NM_001323659.1
NP_001310588.1
7
2562
NM_001323660.1
NP_001310589.1
7
2559
NM_001323661.1
NP_001310590.1
8
2705
NM_001323662.1
NP_001310591.1
9
3107
NM_001323663.1
NP_001310592.1
10
3046
NM_001323664.1
NP_001310593.1
9
3030
NM_001323665.1
NP_001310594.1
8
2948
NM_001323666.1
NP_001310595.1
7
2888
NM_001323671.1
NP_001310600.1
8
2784
NM_001323672.1
NP_001310601.1
9
2836
NM_001323673.1
NP_001310602.1
8
2781
NM_001323674.1
NP_001310603.1
5
2306
NM_001323675.1
NP_001310604.1
5
2416
NM_001323676.1
NP_001310605.1
7
2641
NM_001323677.1
NP_001310606.1
7
2638
NM_001323678.1
NP_001310607.1
5
2583
NM_030751.5
NP_110378.3
7
2528
XM_006717498.2
XP_006717561.1
10
2966
XM_011519643.2
XP_011517945.1
10
3269
XM_017016597.1
XP_016872086.1
10
2868
XM_017016598.1
XP_016872087.1
10
5998
XM_017016599.1
XP_016872088.1
9
5836
XM_017016600.1
XP_016872089.1
11
6077
XM_017016601.1
XP_016872090.1
10
5995
XM_017016602.1
XP_016872091.1
11
6074
XM_017016603.1
XP_016872092.1
10
5912
GenBank mRNA
AK091478.1
-
6
2428
AK294775.1
-
7
2723
AK300830.1
-
6
2275
AK307510.1
-
8
2534
AK314683.1
-
7
2489
AL831979.1
-
9
2786
BC009572.1
-
7
2484
BC112392.1
-
7
2484
40036600
BX647794.1
-
11
3179
M81699.1
-
1
1400
U12170.1
-
7
2489
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0