Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201334.1 | NP_001188263.1 | 3 | 355 | ||
NM_001201335.1 | NP_001188264.1 | 3 | 587 | |||
NM_003203.4 | NP_003194.3 | 3 | 587 | |||
XM_005264520.3 | XP_005264577.1 | 3 | 612 | |||
XM_011533075.1 | XP_011531377.1 | 3 | 612 | |||
XM_017004787.1 | XP_016860276.1 | 3 | 612 | |||
XM_017004788.1 | XP_016860277.1 | 3 | 588 | |||
GenBank mRNA | AB026911.1 | - | 3 | 525 | ||
AK097564.1 | - | 3 | 355 | |||
BC000853.1 | - | 3 | 501 | |||
BC064559.1 | - | 3 | 484 | 5548897 | ||
BU189326.1 | - | 3 | 621 | |||
BU623860.1 | - | 1 | 578 | |||
EF158467.1 | - | 3 | 517 | |||
EF158468.1 | - | 3 | 517 |