Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF12 SIRNA Assays ›
Gene Symbol:
TCF12
Gene Name:
transcription factor 12
Gene Aliases:
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
Chromosome Location:
Chr.15: 56918090 - 57291261 on Build GRCh38
UniGene ID:
Hs.511504
Species:
Human
siRNA ID
HSS110554
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306219.2
NP_001293148.1
10
1081
NM_001306220.2
NP_001293149.1
8
869
NM_001322151.1
NP_001309080.1
17
1788
NM_001322152.1
NP_001309081.1
17
1793
NM_001322154.1
NP_001309083.1
17
1788
NM_001322156.1
NP_001309085.1
18
1855
NM_001322157.1
NP_001309086.1
16
1721
NM_001322158.1
NP_001309087.1
17
1783
NM_001322159.1
NP_001309088.1
17
1793
NM_001322161.1
NP_001309090.1
17
1636
NM_001322162.1
NP_001309091.1
17
1639
NM_001322164.1
NP_001309093.1
16
1586
NM_001322165.1
NP_001309094.1
15
1550
NM_003205.3
NP_003196.1
16
1721
NM_207036.1
NP_996919.1
17
1783
NM_207037.1
NP_996920.1
17
1793
NM_207038.1
NP_996921.1
16
1711
NM_207040.1
NP_996923.1
9
982
XM_011521959.2
XP_011520261.1
18
1789
XM_011521960.2
XP_011520262.1
18
1789
XM_011521961.2
XP_011520263.1
18
1789
XM_011521962.2
XP_011520264.1
17
1717
XM_011521963.2
XP_011520265.1
17
1717
XM_011521965.1
XP_011520267.1
16
1500
XM_011521966.2
XP_011520268.1
9
1396
XM_011521967.1
XP_011520269.1
15
1428
XM_011521969.1
XP_011520271.1
9
975
XM_017022520.1
XP_016878009.1
16
1687
GenBank mRNA
AK294617.1
-
17
1741
AK294991.1
-
10
1081
AK302749.1
-
5
427
AK302923.1
-
16
1671
AK303028.1
-
4
379
AK304007.1
-
8
864
AK308101.1
-
10
1055
AK311709.1
-
3
533
AK312710.1
-
17
1753
AL831981.2
-
16
1689
BC050556.1
-
17
1734
5767579
BX537967.1
-
9
973
M65209.1
-
4
374
M80627.1
-
16
1640
M83233.1
-
16
1515
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
immune response
muscle organ development
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
enhancer binding
cAMP response element binding
protein homodimerization activity
bHLH transcription factor binding
transcription regulatory region DNA binding
SMAD binding
protein heterodimerization activity
E-box binding
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0