Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005651.3 | NP_005642.1 | 8 | 865 | ||
GenBank mRNA | AK289418.1 | - | 8 | 862 | ||
AK307741.1 | - | 8 | 862 | |||
BC005355.1 | - | 8 | 862 | 4071714 | ||
BX647341.1 | - | 5 | 2308 | |||
U32989.1 | - | 8 | 863 |