Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021961.5 | NP_068780.2 | 11 | 1585 | ||
GenBank mRNA | AB209858.1 | - | 6 | 900 | ||
AL833289.1 | - | 12 | 1194 | |||
BC115398.1 | - | 10 | 916 | 40068609 |