Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021961.5 | NP_068780.2 | 5, 6 | 950 | ||
GenBank mRNA | AL035973.1 | - | 3, 4 | 235 |