Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THRB SIRNA Assays ›
Gene Symbol:
THRB
Gene Name:
thyroid hormone receptor beta
Gene Aliases:
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
Chromosome Location:
Chr.3: 24117153 - 24495281 on Build GRCh38
UniGene ID:
Hs.187861
Species:
Human
siRNA ID
HSS110743
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THRB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000461.4
NP_000452.2
4
386
NM_001128176.2
NP_001121648.1
5
497
NM_001128177.1
NP_001121649.1
5
523
NM_001252634.1
NP_001239563.1
6
621
XM_005265421.4
XP_005265478.1
5
594
XM_005265423.4
XP_005265480.1
4
429
XM_005265424.3
XP_005265481.1
4
364
XM_006713317.3
XP_006713380.1
5
737
XM_006713318.3
XP_006713381.1
5
689
XM_011534046.2
XP_011532348.1
5
2637
XM_011534047.2
XP_011532349.1
6
874
XM_011534048.2
XP_011532350.1
5
453
XM_011534049.2
XP_011532351.1
4
309
XM_011534050.2
XP_011532352.1
5
682
XM_011534051.2
XP_011532353.1
4
272
XM_011534052.2
XP_011532354.1
5
409
XM_011534053.2
XP_011532355.1
4
530
XM_017007108.1
XP_016862597.1
6
807
XM_017007109.1
XP_016862598.1
7
965
XM_017007110.1
XP_016862599.1
6
772
XM_017007111.1
XP_016862600.1
5
418
XM_017007112.1
XP_016862601.1
4
388
XM_017007113.1
XP_016862602.1
3
527
XM_017007114.1
XP_016862603.1
3
492
XM_017007115.1
XP_016862604.1
3
366
GenBank mRNA
AB307687.1
-
1
86
AK096628.1
-
6
610
AY286465.1
-
4
312
AY286466.1
-
4
330
AY286467.1
-
4
296
AY286468.1
-
5
415
AY286469.1
-
5
406
AY286470.1
-
5
440
AY286471.1
-
6
489
BC106929.1
-
3
211
40033198
BC106930.1
-
4
357
40033200
BF854291.1
-
1
323
BP313943.1
-
4
375
BQ879477.1
-
4
508
DA218319.1
-
4
326
GQ919288.1
-
4
249
HQ692825.1
-
2
101
M26747.1
-
4
386
X04707.1
-
4
386
Back To Top
More Information
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
organ morphogenesis
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
Type I pneumocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription corepressor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
enzyme binding
chromatin DNA binding
sequence-specific DNA binding
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0