Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE4 SIRNA Assays ›
Gene Symbol:
TLE4
Gene Name:
transducin like enhancer of split 4
Gene Aliases:
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
Chromosome Location:
Chr.9: 79571773 - 79726882 on Build GRCh38
UniGene ID:
Hs.444213
Species:
Human
siRNA ID
HSS110794
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282748.1
NP_001269677.1
7
1430
NM_001282749.1
NP_001269678.1
6
1355
NM_001282753.1
NP_001269682.1
7
1430
NM_001282760.1
NP_001269689.1
8
1525
NM_007005.4
NP_008936.2
7
1430
NR_104239.1
7
1433
XM_005252177.1
XP_005252234.1
7
606
XM_006717264.1
XP_006717327.1
9
729
XM_006717268.1
XP_006717331.1
9
729
XM_011518952.1
XP_011517254.1
9
729
XM_011518953.1
XP_011517255.1
9
606
XM_011518954.1
XP_011517256.1
8
684
XM_011518955.1
XP_011517257.1
8
666
XM_011518956.1
XP_011517258.1
8
645
XM_011518957.1
XP_011517259.1
8
642
XM_011518958.1
XP_011517260.1
9
729
XM_011518959.1
XP_011517261.1
7
606
XM_011518960.1
XP_011517262.1
7
603
XM_011518961.1
XP_011517263.1
6
543
XM_011518962.1
XP_011517264.1
6
540
XM_011518963.1
XP_011517265.1
6
531
XM_011518964.1
XP_011517266.1
6
528
XM_011518965.1
XP_011517267.1
7
606
XM_011518966.1
XP_011517268.1
7
603
XM_011518967.1
XP_011517269.1
9
729
XM_011518968.1
XP_011517270.1
5
468
XM_011518969.1
XP_011517271.1
9
729
XM_011518970.2
XP_011517272.1
9
729
XM_017015067.1
XP_016870556.1
7
534
XM_017015068.1
XP_016870557.1
6
561
XM_017015069.1
XP_016870558.1
6
389
XM_017015070.1
XP_016870559.1
6
466
XM_017015071.1
XP_016870560.1
8
642
XM_017015072.1
XP_016870561.1
7
563
XM_017015073.1
XP_016870562.1
7
565
XM_017015074.1
XP_016870563.1
9
729
XM_017015075.1
XP_016870564.1
6
558
XM_017015076.1
XP_016870565.1
6
505
XM_017015077.1
XP_016870566.1
6
543
XM_017015078.1
XP_016870567.1
6
540
XM_017015079.1
XP_016870568.1
6
468
XM_017015080.1
XP_016870569.1
6
531
XM_017015081.1
XP_016870570.1
7
606
XM_017015082.1
XP_016870571.1
6
423
XM_017015083.1
XP_016870572.1
7
603
XM_017015084.1
XP_016870573.1
6
528
XM_017015085.1
XP_016870574.1
6
436
XM_017015086.1
XP_016870575.1
6
528
GenBank mRNA
AB033087.1
-
7
491
AK057236.1
-
7
581
AK296342.1
-
6
384
AK302709.1
-
7
465
AL162059.1
-
5
265
BC036369.1
-
8
725
BC045650.1
-
7
1430
5268902
BC059405.1
-
7
588
5296117
HQ283388.1
-
44
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
biological_process
Wnt signaling pathway
beta-catenin-TCF complex assembly
molecular_function
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0