Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE4 SIRNA Assays ›
Gene Symbol:
TLE4
Gene Name:
transducin like enhancer of split 4
Gene Aliases:
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
Chromosome Location:
Chr.9: 79571773 - 79726882 on Build GRCh38
UniGene ID:
Hs.444213
Species:
Human
siRNA ID
HSS110796
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282748.1
NP_001269677.1
15
2404
NM_001282749.1
NP_001269678.1
13
2233
NM_001282753.1
NP_001269682.1
12
2101
NM_001282760.1
NP_001269689.1
15
2403
NM_007005.4
NP_008936.2
14
2308
NR_104239.1
15
2441
XM_005252177.1
XP_005252234.1
14
1484
XM_006717264.1
XP_006717327.1
17
1703
XM_006717268.1
XP_006717331.1
16
1607
XM_011518952.1
XP_011517254.1
17
1703
XM_011518953.1
XP_011517255.1
17
1580
XM_011518954.1
XP_011517256.1
16
1658
XM_011518955.1
XP_011517257.1
16
1640
XM_011518956.1
XP_011517258.1
16
1619
XM_011518957.1
XP_011517259.1
16
1616
XM_011518958.1
XP_011517260.1
16
1607
XM_011518959.1
XP_011517261.1
15
1580
XM_011518960.1
XP_011517262.1
15
1577
XM_011518961.1
XP_011517263.1
14
1517
XM_011518962.1
XP_011517264.1
14
1514
XM_011518963.1
XP_011517265.1
14
1505
XM_011518964.1
XP_011517266.1
14
1502
XM_011518965.1
XP_011517267.1
14
1484
XM_011518966.1
XP_011517268.1
14
1481
XM_011518967.1
XP_011517269.1
15
1454
XM_011518968.1
XP_011517270.1
13
1442
XM_011518969.1
XP_011517271.1
14
1400
XM_011518970.2
XP_011517272.1
17
1703
XM_011518972.2
XP_011517274.1
5
530
XM_017015067.1
XP_016870556.1
15
1508
XM_017015068.1
XP_016870557.1
14
1535
XM_017015069.1
XP_016870558.1
14
1363
XM_017015070.1
XP_016870559.1
14
1440
XM_017015071.1
XP_016870560.1
15
1520
XM_017015072.1
XP_016870561.1
14
1441
XM_017015073.1
XP_016870562.1
14
1443
XM_017015074.1
XP_016870563.1
15
1454
XM_017015075.1
XP_016870564.1
13
1436
XM_017015076.1
XP_016870565.1
13
1383
XM_017015077.1
XP_016870566.1
13
1421
XM_017015078.1
XP_016870567.1
13
1418
XM_017015079.1
XP_016870568.1
13
1346
XM_017015080.1
XP_016870569.1
13
1409
XM_017015081.1
XP_016870570.1
13
1331
XM_017015082.1
XP_016870571.1
13
1301
XM_017015083.1
XP_016870572.1
13
1328
XM_017015084.1
XP_016870573.1
12
1253
XM_017015085.1
XP_016870574.1
12
1161
XM_017015086.1
XP_016870575.1
11
1199
GenBank mRNA
AB033087.1
-
14
1369
AK057236.1
-
15
1589
AK296342.1
-
13
1262
AL162059.1
-
12
1143
BC036369.1
-
15
1603
BC045650.1
-
12
2101
5268902
BC059405.1
-
15
1562
5296117
M99439.1
-
3
315
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
biological_process
Wnt signaling pathway
beta-catenin-TCF complex assembly
molecular_function
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0