Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007118.3 | NP_009049.2 | 53 | 8282 | ||
NR_134469.1 | 53 | 7752 | ||||
XM_011514107.2 | XP_011512409.1 | 53 | 8857 | |||
XM_011514108.1 | XP_011512410.1 | 53 | 8156 | |||
XM_011514109.2 | XP_011512411.1 | 53 | 11934 | |||
XM_011514110.2 | XP_011512412.1 | 53 | 8392 | |||
XM_017009801.1 | XP_016865290.1 | 53 | 8622 | |||
XM_017009802.1 | XP_016865291.1 | 53 | 8634 | |||
XM_017009803.1 | XP_016865292.1 | 46 | 6903 | |||
GenBank mRNA | AF091395.1 | - | 53 | 8147 | ||
AK131423.1 | - | 6 | 1248 | |||
AL161955.1 | - | 2 | 1324 | |||
BC017268.2 | - | 2 | 118 | |||
U42390.1 | - | 53 | 7616 |