Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152653.3 | NP_689866.1 | 5 | 596 | ||
XM_005265432.2 | XP_005265489.1 | 5 | 680 | |||
XM_005265433.4 | XP_005265490.1 | 5 | 680 | |||
GenBank mRNA | AK057886.1 | - | 5 | 597 | ||
AK307075.1 | - | 4 | 392 | |||
BC022332.1 | - | 5 | 572 | 4295679 |