Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003365.2 | NP_003356.2 | 6, 7 | 762 | ||
GenBank mRNA | AK129903.1 | - | 6, 7 | 742 | ||
AK300694.1 | - | 6, 7 | 697 | |||
AK313090.1 | - | 6, 7 | 732 | |||
BC009586.1 | - | 6, 7 | 733 | 4096628 | ||
D26485.1 | - | 6, 7 | 724 | |||
L16842.1 | - | 6, 7 | 1134 |