Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111077.1 | NP_001104547.1 | 4 | 352 | ||
NM_003379.4 | NP_003370.2 | 3 | 386 | |||
GenBank mRNA | AK296738.1 | - | 4 | 315 | ||
AK312597.1 | - | 4 | 296 | |||
AL162086.1 | - | 4 | 304 | |||
BC013903.2 | - | 3 | 303 | 2959399 | ||
BC068458.1 | - | 3 | 312 | 4819793 | ||
DA754543.1 | - | 4 | 352 | |||
FJ224318.1 | - | 3 | 288 | |||
J05021.1 | - | 3 | 247 | |||
X51521.1 | - | 3 | 288 |