Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251882.1 | NP_001238811.1 | 4, 5 | 791 | ||
NM_001251883.1 | NP_001238812.1 | 2, 3 | 308 | |||
NM_001251884.1 | NP_001238813.1 | 2, 3 | 308 | |||
NM_001251885.1 | NP_001238814.1 | 3, 4 | 368 | |||
NM_004624.3 | NP_004615.2 | 3, 4 | 414 | |||
XM_005265437.2 | XP_005265494.1 | 3, 4 | 383 | |||
XM_005265438.3 | XP_005265495.1 | 3, 4 | 943 | |||
XM_005265439.2 | XP_005265496.1 | 2, 3 | 277 | |||
XM_011534079.1 | XP_011532381.1 | 4, 5 | 1142 | |||
XM_011534080.2 | XP_011532382.1 | 3, 4 | 397 | |||
GenBank mRNA | AK056819.1 | - | 4, 5 | 791 | ||
AK293548.1 | - | 2, 3 | 303 | |||
AK294609.1 | - | 2, 3 | 308 | |||
AK298115.1 | - | 3, 4 | 337 | |||
AK314334.1 | - | 3, 4 | 383 | |||
BC064424.1 | - | 3, 4 | 370 | 5752796 | ||
DA200439.1 | - | 2, 3 | 308 | |||
EF577396.1 | - | 3, 4 | 290 | |||
L13288.1 | - | 3, 4 | 384 | |||
L20295.1 | - | 2, 3 | 194 | |||
X75299.1 | - | 3, 4 | 301 |
Set Membership: |
![]() |