Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005433.3 | NP_005424.1 | 7 | 1052 | ||
XM_005258139.3 | XP_005258196.1 | 7 | 1014 | |||
XM_017025960.1 | XP_016881449.1 | 7 | 894 | |||
GenBank mRNA | AK314063.1 | - | 7 | 996 | ||
BC048960.1 | - | 7 | 1048 | 5260751 | ||
M15990.1 | - | 7 | 1038 |