Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFX SIRNA Assays ›
Gene Symbol:
ZFX
Gene Name:
zinc finger protein, X-linked
Gene Aliases:
ZNF926
Chromosome Location:
Chr.X: 24148994 - 24216255 on Build GRCh38
UniGene ID:
Hs.336681
Species:
Human
siRNA ID
HSS111455
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178084.1
NP_001171555.1
8
1688
NM_001178085.1
NP_001171556.1
11
2032
NM_001178086.1
NP_001171557.1
6
1093
NM_001178095.1
NP_001171566.1
8
1826
NM_003410.3
NP_003401.2
10
1896
XM_005274591.4
XP_005274648.1
9
2062
XM_005274592.3
XP_005274649.1
8
1877
XM_005274594.4
XP_005274651.1
7
3067
XM_006724513.3
XP_006724576.1
11
2291
XM_006724515.3
XP_006724578.1
10
2152
XM_011545578.1
XP_011543880.1
11
2173
XM_011545579.1
XP_011543881.1
10
1961
XM_011545581.2
XP_011543883.1
10
2103
XM_011545583.2
XP_011543885.1
7
1617
XM_017029788.1
XP_016885277.1
11
2214
XM_017029789.1
XP_016885278.1
10
2776
XM_017029790.1
XP_016885279.1
10
2181
XM_017029791.1
XP_016885280.1
9
2071
XM_017029792.1
XP_016885281.1
9
1950
XM_017029793.1
XP_016885282.1
8
1748
XM_017029794.1
XP_016885283.1
10
2002
XM_017029795.1
XP_016885284.1
7
1631
XM_017029796.1
XP_016885285.1
8
1642
XM_017029797.1
XP_016885286.1
11
2242
XM_017029798.1
XP_016885287.1
7
1862
XM_017029799.1
XP_016885288.1
7
1736
XM_017029800.1
XP_016885289.1
6
1723
XM_017029801.1
XP_016885290.1
6
1480
GenBank mRNA
AB209892.1
-
8
1859
AF454950.1
-
4
803
AY072770.1
-
3
650
BC136308.1
-
10
1884
9020295
BC136309.1
-
10
1884
9020296
M30608.1
-
9
1847
X59738.1
-
6
1090
X59739.1
-
8
1664
X59740.1
-
11
2013
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0