Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305033.1 | NP_001291962.1 | 4 | 2012 | ||
NM_001305035.1 | NP_001291964.1 | 6 | 2142 | |||
NM_001305036.1 | NP_001291965.1 | 5 | 2070 | |||
NM_001305037.1 | NP_001291966.1 | 5 | 1997 | |||
NM_001305038.1 | NP_001291967.1 | 5 | 2089 | |||
NM_001305039.1 | NP_001291968.1 | 6 | 2147 | |||
NM_001305040.1 | NP_001291969.1 | 7 | 2223 | |||
NM_006955.2 | NP_008886.1 | 5 | 2065 | |||
GenBank mRNA | AJ491697.1 | - | 5 | 1772 | ||
AK025638.1 | - | 1 | 1146 | |||
AK304306.1 | - | 5 | 1696 | |||
BC048313.2 | - | 5 | 1770 | |||
BC107726.1 | - | 5 | 1763 |