Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185063.2 | NP_001171992.1 | 2 | 549 | ||
NM_007131.4 | NP_009062.2 | 3 | 2869 | |||
XM_005262469.1 | XP_005262526.1 | 3 | 1398 | |||
XM_005262471.3 | XP_005262528.1 | 1 | 1780 | |||
XM_011531391.1 | XP_011529693.1 | 4 | 1585 | |||
XM_017029818.1 | XP_016885307.1 | 3 | 2891 | |||
XM_017029819.1 | XP_016885308.1 | 2 | 4400 | |||
GenBank mRNA | AK091687.1 | - | 2 | 543 | ||
AK290215.1 | - | 2 | 573 | |||
AL831984.2 | - | 3 | 2814 | |||
AL832801.1 | - | 2 | 356 | |||
BC109100.2 | - | 2 | 388 | 40034784 |