Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185063.2 | NP_001171992.1 | 2 | 772 | ||
NM_007131.4 | NP_009062.2 | 3 | 3092 | |||
XM_005262469.1 | XP_005262526.1 | 3 | 1621 | |||
XM_005262471.3 | XP_005262528.1 | 1 | 2003 | |||
XM_011531391.1 | XP_011529693.1 | 4 | 1808 | |||
XM_017029818.1 | XP_016885307.1 | 3 | 3114 | |||
XM_017029819.1 | XP_016885308.1 | 2 | 4623 | |||
GenBank mRNA | AK091687.1 | - | 2 | 766 | ||
AK290215.1 | - | 2 | 796 | |||
AL831984.2 | - | 3 | 3037 | |||
AL832801.1 | - | 2 | 579 | |||
BC109100.2 | - | 2 | 611 | 40034784 |