Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297588.1 | NP_001284517.1 | 5 | 1897 | ||
NM_001297589.1 | NP_001284518.1 | 5 | 1907 | |||
NM_013359.2 | NP_037491.2 | 6 | 2045 | |||
XM_017027232.1 | XP_016882721.1 | 5 | 1924 | |||
XM_017027233.1 | XP_016882722.1 | 4 | 1841 | |||
GenBank mRNA | AF187987.1 | - | 6 | 2045 | ||
AK314207.1 | - | 5 | 1892 | |||
BC111981.1 | - | 5 | 1800 | 8327449 | ||
BC113426.1 | - | 5 | 1800 | 8322478 | ||
BC143659.1 | - | 6 | 1948 |
Gene Symbol: ZNF155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527279.2 | XP_011525581.1 | 1 | 214 |