Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007153.3 | NP_009084.2 | 4 | 678 | ||
XM_017027257.1 | XP_016882746.1 | 4 | 725 |