Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF227 SIRNA Assays ›
Gene Symbol:
ZNF227
Gene Name:
zinc finger protein 227
Gene Aliases:
-
Chromosome Location:
Chr.19: 44207527 - 44237268 on Build GRCh38
UniGene ID:
Hs.298089
Species:
Human
siRNA ID
HSS111704
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF227
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289166.1
NP_001276095.1
6
693
NM_001289167.1
NP_001276096.1
6
649
NM_001289168.1
NP_001276097.1
5
628
NM_001289169.1
NP_001276098.1
5
631
NM_001289170.1
NP_001276099.1
4
544
NM_001289171.1
NP_001276100.1
4
547
NM_001289172.1
NP_001276101.1
4
605
NM_001289173.1
NP_001276102.1
5
627
NM_182490.2
NP_872296.1
6
690
XM_005259232.2
XP_005259289.1
6
821
XM_006723371.1
XP_006723434.1
5
602
XM_011527289.2
XP_011525591.1
6
969
XM_011527290.1
XP_011525592.1
6
665
XM_017027266.1
XP_016882755.1
6
824
XM_017027267.1
XP_016882756.1
5
705
XM_017027268.1
XP_016882757.1
5
759
XM_017027269.1
XP_016882758.1
5
729
XM_017027270.1
XP_016882759.1
5
907
XM_017027271.1
XP_016882760.1
5
905
XM_017027272.1
XP_016882761.1
5
570
XM_017027273.1
XP_016882762.1
5
644
XM_017027274.1
XP_016882763.1
5
909
XM_017027275.1
XP_016882764.1
6
792
XM_017027276.1
XP_016882765.1
6
788
XM_017027277.1
XP_016882766.1
5
652
XM_017027278.1
XP_016882767.1
3
286
XM_017027279.1
XP_016882768.1
4
678
GenBank mRNA
AK092253.1
-
6
690
AK299266.1
-
4
581
AK300576.1
-
4
502
AK316048.1
-
6
639
AL833012.1
-
4
448
AY166786.1
-
3
340
BC047570.1
-
6
680
4838846
BU153285.1
-
4
583
DC341721.1
-
4
507
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0