Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC30A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003459.4 | NP_003450.2 | 1, 2 | 273 | ||
XM_005264547.1 | XP_005264604.1 | 1, 2 | 269 | |||
XM_011533103.2 | XP_011531405.1 | 1, 2 | 261 | |||
GenBank mRNA | AK313600.1 | - | 1, 2 | 228 | ||
AL043136.2 | - | 1, 2 | 273 | |||
BC028358.2 | - | 1, 2 | 261 | 4837595 | ||
U76010.1 | - | 1, 2 | 170 |