Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013309.5 | NP_037441.2 | 4 | 984 | ||
XM_011521997.2 | XP_011520299.1 | 4 | 1488 | |||
XM_017022560.1 | XP_016878049.1 | 3 | 790 | |||
GenBank mRNA | AF025409.1 | - | 3 | 672 | ||
AK290874.1 | - | 3 | 790 | |||
BC026089.2 | - | 4 | 984 | 4829839 |