Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057176.2 | NP_476517.1 | 1 | 265 | ||
GenBank mRNA | AK129999.1 | - | 1 | 276 | ||
AY034632.1 | - | 1 | 265 | |||
BC103898.1 | - | 1 | 244 | 40005137 | ||
BC103899.1 | - | 1 | 244 | 40005138 | ||
BC103900.1 | - | 1 | 244 | 40005139 |