Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006763.2 | NP_006754.1 | 2 | 533 | ||
GenBank mRNA | BC105948.1 | - | 2 | 507 | 40032676 | |
BC105949.1 | - | 2 | 507 | 40032675 | ||
U72649.1 | - | 2 | 533 | |||
Y09943.1 | - | 2 | 525 |