Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015120.4 | NP_055935.4 | 12 | 9932 | ||
GenBank mRNA | AB002326.3 | - | 5 | 3621 | ||
AJ417593.2 | - | 12 | 9932 | |||
AL833495.1 | - | 5 | 2764 | |||
AX751581.1 | - | 11 | 9806 | |||
BC035025.2 | - | 5 | 2620 | |||
BC050330.1 | - | 5 | 2620 | |||
BX647706.1 | - | 5 | 3845 | |||
BX648632.1 | - | 5 | 4345 | |||
EF151141.1 | - | 2 | 109 |