Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146158.1 | NP_001139630.1 | 4, 5 | 610 | ||
NM_006302.2 | NP_006293.2 | 3, 4 | 936 | |||
XM_017004876.1 | XP_016860365.1 | 2, 3 | 699 | |||
XM_017004877.1 | XP_016860366.1 | 3, 4 | 374 | |||
GenBank mRNA | AK292553.1 | - | 3, 4 | 882 | ||
AK292719.1 | - | 1, 2 | 454 | |||
AK307991.1 | - | 4, 5 | 596 | |||
AK313355.1 | - | 3, 4 | 936 | |||
BC028337.1 | - | 3, 4 | 917 | 5110525 | ||
BC117250.1 | - | 3, 4 | 809 | 40125801 | ||
BC117252.1 | - | 3, 4 | 809 | 40125803 | ||
DA520534.1 | - | 2, 3 | 291 | |||
X87237.1 | - | 3, 4 | 905 |