Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAG6 SIRNA Assays ›
Gene Symbol:
BAG6
Gene Name:
BCL2 associated athanogene 6
Gene Aliases:
BAG-6, BAT3, D6S52E, G3
Chromosome Location:
Chr.6: 31639028 - 31660900 on Build GRCh38
UniGene ID:
Hs.440900
Species:
Human
siRNA ID
HSS111844
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAG6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098534.1
NP_001092004.1
10
1514
NM_001199697.1
NP_001186626.1
10
1644
NM_001199698.1
NP_001186627.1
10
1644
NM_004639.3
NP_004630.3
10
1686
NM_080702.2
NP_542433.1
10
1644
NM_080703.2
NP_542434.1
10
1668
XM_011514892.2
XP_011513194.1
10
2627
XM_017011275.1
XP_016866764.1
10
2627
XM_017011276.1
XP_016866765.1
10
1617
XM_017011277.1
XP_016866766.1
10
1477
XM_017011278.1
XP_016866767.1
10
1552
XM_017011279.1
XP_016866768.1
10
1947
XM_017011280.1
XP_016866769.1
10
1519
XM_017011281.1
XP_016866770.1
10
1681
XM_017011282.1
XP_016866771.1
10
1473
XM_017011283.1
XP_016866772.1
10
2627
XM_017011284.1
XP_016866773.1
10
2627
XM_017011285.1
XP_016866774.1
9
1417
XM_017011286.1
XP_016866775.1
9
1436
XM_017011287.1
XP_016866776.1
10
2627
XM_017011288.1
XP_016866777.1
9
1417
XM_017011289.1
XP_016866778.1
10
2627
XM_017011290.1
XP_016866779.1
10
2627
XM_017011291.1
XP_016866780.1
10
2627
XM_017011292.1
XP_016866781.1
10
2627
XM_017011293.1
XP_016866782.1
9
1417
XM_017011294.1
XP_016866783.1
10
2627
XM_017011295.1
XP_016866784.1
10
2627
XM_017011296.1
XP_016866785.1
10
2627
XM_017011297.1
XP_016866786.1
9
1417
XM_017011298.1
XP_016866787.1
10
2627
GenBank mRNA
AK302695.1
-
10
1595
AK304879.1
-
10
1595
BC003133.1
-
10
1562
2957896
BX647244.1
-
10
1522
M33519.1
-
10
1621
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
immune system process
ubiquitin-dependent protein catabolic process
transport
synaptonemal complex assembly
spermatogenesis
brain development
embryo development
covalent chromatin modification
internal peptidyl-lysine acetylation
cell differentiation
lung development
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of apoptotic process
negative regulation of proteolysis
protein stabilization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
tail-anchored membrane protein insertion into ER membrane
positive regulation of ERAD pathway
maintenance of unfolded protein involved in ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
protein binding
Hsp70 protein binding
polyubiquitin binding
ubiquitin protein ligase binding
ribosome binding
misfolded protein binding
proteasome binding
ubiquitin-specific protease binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline