Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167827.2 | NP_001161299.2 | 7 | 907 | ||
NM_001271943.1 | NP_001258872.1 | 6 | 716 | |||
NM_001271944.1 | NP_001258873.1 | 7 | 971 | |||
NM_001271945.1 | NP_001258874.1 | 7 | 882 | |||
NM_007162.2 | NP_009093.1 | 8 | 1073 | |||
XM_005249411.1 | XP_005249468.1 | 7 | 1006 | |||
XM_005249412.1 | XP_005249469.1 | 7 | 898 | |||
XM_006715212.3 | XP_006715275.1 | 7 | 1125 | |||
XM_006715213.2 | XP_006715276.1 | 7 | 949 | |||
XM_011514915.1 | XP_011513217.1 | 7 | 1128 | |||
XM_011514916.2 | XP_011513218.1 | 8 | 918 | |||
GenBank mRNA | AJ608786.1 | - | 7 | 907 | ||
AK095061.1 | - | 7 | 1998 | |||
AK298390.1 | - | 7 | 892 | |||
BC006225.2 | - | 5 | 345 | |||
BC032448.1 | - | 8 | 1073 | 5180066 | ||
CT000098.1 | - | 6 | 700 | |||
M33782.1 | - | 7 | 1053 |