Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324282.1 | NP_001311211.1 | 11 | 1227 | ||
NM_001324283.1 | NP_001311212.1 | 11 | 1238 | |||
NM_001324284.1 | NP_001311213.1 | 13 | 1427 | |||
NM_001324285.1 | NP_001311214.1 | 13 | 1435 | |||
NM_001324286.1 | NP_001311215.1 | 9 | 1066 | |||
NM_001324287.1 | NP_001311216.1 | 10 | 1151 | |||
NM_001324288.1 | NP_001311217.1 | 13 | 1466 | |||
NM_001324289.1 | NP_001311218.1 | 12 | 1381 | |||
NM_003473.3 | NP_003464.1 | 12 | 1323 | |||
XM_011519695.2 | XP_011517997.1 | 12 | 1299 | |||
XM_017016685.1 | XP_016872174.1 | 8 | 1368 | |||
GenBank mRNA | AK303083.1 | - | 10 | 1011 | ||
AK314405.1 | - | 12 | 1207 | |||
BC030586.2 | - | 12 | 1266 | 4820857 | ||
U43899.1 | - | 12 | 1150 |