Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195626.1 | NP_001182555.1 | 1 | 68 | ||
NM_001195627.1 | NP_001182556.1 | 2 | 417 | |||
NM_001195628.1 | NP_001182557.1 | 2 | 391 | |||
NM_001195630.1 | NP_001182559.1 | 2 | 417 | |||
NM_001324296.1 | NP_001311225.1 | 2 | 561 | |||
NM_001324297.1 | NP_001311226.1 | 2 | 561 | |||
NM_004641.3 | NP_004632.1 | 2 | 417 | |||
NR_136736.1 | 2 | 391 | ||||
GenBank mRNA | AB209755.1 | - | 2 | 97 | ||
AY598745.1 | - | 1 | 68 | |||
BC065376.1 | - | 2 | 246 | |||
BC080577.1 | - | 2 | 246 | |||
BC129946.1 | - | 2 | 110 | |||
U13948.1 | - | 2 | 251 |