Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CUL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003478.3 | NP_003469.2 | 18, 19 | 2821 | ||
XM_005271682.2 | XP_005271739.1 | 17, 18 | 2428 | |||
XM_017018364.1 | XP_016873853.1 | 19, 20 | 2299 | |||
XM_017018365.1 | XP_016873854.1 | 19, 20 | 2315 | |||
GenBank mRNA | AF017061.1 | - | 18, 19 | 2157 | ||
AF327710.1 | - | 18, 19 | 2154 | |||
AK125294.1 | - | 18, 19 | 2770 | |||
AK292575.1 | - | 18, 19 | 2584 | |||
BC063306.1 | - | 18, 19 | 2584 | 30331132 | ||
X81882.1 | - | 18, 19 | 2243 |