Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242902.1 | NP_001229831.1 | 11 | 2877 | ||
NM_001242903.1 | NP_001229832.1 | 11 | 3150 | |||
NM_003488.3 | NP_003479.1 | 10 | 2826 | |||
XM_005257707.2 | XP_005257764.1 | 11 | 3146 | |||
XM_005257709.2 | XP_005257766.1 | 10 | 2938 | |||
XM_017025189.1 | XP_016880678.1 | 11 | 2884 | |||
XM_017025190.1 | XP_016880679.1 | 10 | 3047 | |||
XM_017025191.1 | XP_016880680.1 | 10 | 2683 | |||
GenBank mRNA | AK292416.1 | - | 10 | 2805 | ||
AK297810.1 | - | 11 | 2876 | |||
AK308397.1 | - | 11 | 2959 | |||
BC000729.2 | - | 10 | 2758 | 3509111 | ||
CD172211.1 | - | 3 | 125 | |||
X97335.1 | - | 10 | 2719 |