Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF239 SIRNA Assays ›
Gene Symbol:
ZNF239
Gene Name:
zinc finger protein 239
Gene Aliases:
HOK-2, MOK2
Chromosome Location:
Chr.10: 43554516 - 43574619 on Build GRCh38
UniGene ID:
Hs.25040
Species:
Human
siRNA ID
HSS112029
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF239
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099282.1
NP_001092752.1
4
489
NM_001099283.1
NP_001092753.1
2
326
NM_001099284.1
NP_001092754.1
3
449
NM_001324347.1
NP_001311276.1
3
367
NM_001324348.1
NP_001311277.1
2
395
NM_001324349.1
NP_001311278.1
4
559
NM_001324350.1
NP_001311279.1
3
436
NM_001324351.1
NP_001311280.1
3
518
NM_001324352.1
NP_001311281.1
4
494
NM_001324353.1
NP_001311282.1
5
599
NM_005674.2
NP_005665.2
2
808
XM_005271832.2
XP_005271889.1
2
321
XM_006718001.2
XP_006718064.1
2
420
XM_006718003.3
XP_006718066.1
2
320
XM_011540232.2
XP_011538534.1
3
3407
XM_011540234.2
XP_011538536.1
3
454
XM_011540235.2
XP_011538537.1
3
453
XM_011540236.2
XP_011538538.1
3
462
XM_011540237.2
XP_011538539.1
3
412
XM_011540238.2
XP_011538540.1
2
264
XM_017016740.1
XP_016872229.1
4
491
GenBank mRNA
AK292013.1
-
3
445
BC026030.1
-
4
433
5460088
CK820210.1
-
5
541
DA686973.1
-
2
322
DA937379.1
-
3
445
DN994387.1
-
3
494
X82125.1
-
2
808
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0