Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003502.3 | NP_003493.1 | 2 | 1085 | ||
NM_181050.2 | NP_851393.1 | 2 | 1085 | |||
NR_134879.1 | 2 | 1085 | ||||
XM_011522682.2 | XP_011520984.1 | 2 | 1096 | |||
XM_011522683.2 | XP_011520985.1 | 2 | 1095 | |||
XM_011522684.2 | XP_011520986.1 | 2 | 946 | |||
XM_017023743.1 | XP_016879232.1 | 2 | 1096 | |||
XM_017023744.1 | XP_016879233.1 | 2 | 1095 | |||
XM_017023747.1 | XP_016879236.1 | 2 | 1098 | |||
XM_017023748.1 | XP_016879237.1 | 2 | 1098 | |||
GenBank mRNA | AF009674.1 | - | 2 | 808 | ||
BC035872.1 | - | 1 | 126 | |||
BC044648.1 | - | 2 | 857 | 5809104 | ||
KF615850.1 | - | 2 | 1085 | |||
KF615851.1 | - | 2 | 1085 | |||
KF615852.1 | - | 2 | 1085 | |||
KU178413.1 | - | 1 | 696 | |||
KU178414.1 | - | 1 | 696 | |||
KU178415.1 | - | 1 | 696 |