Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003508.2 | NP_003499.1 | 1 | 1954 | ||
GenBank mRNA | BC026333.1 | - | 1 | 1950 | 4791657 | |
U82169.1 | - | 1 | 1750 |
Set Membership: |
![]() |