Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003649.2 | NP_003640.2 | 2 | 179 | ||
NM_004032.2 | NP_004023.2 | 2 | 179 | |||
GenBank mRNA | AB496420.1 | - | 1 | 91 | ||
AK293029.1 | - | 2 | 185 | |||
AX786391.1 | - | 1 | 91 | |||
AX786392.1 | - | 91 | ||||
BC032786.1 | - | 2 | 179 | 5179648 | ||
D89858.1 | - | 1 | 91 |