Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006769.3 | NP_006760.1 | 4 | 1162 | ||
XM_005271291.3 | XP_005271348.1 | 4 | 3332 | |||
GenBank mRNA | AK292578.1 | - | 4 | 764 | ||
BC003600.1 | - | 4 | 525 | 3355972 | ||
BC017673.1 | - | 4 | 650 | 3893112 | ||
BC065818.1 | - | 4 | 456 | 6196307 | ||
U24576.1 | - | 4 | 1162 |