Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006769.3 | NP_006760.1 | 4, 5 | 1264 | ||
XM_005271291.3 | XP_005271348.1 | 4, 5 | 3434 | |||
GenBank mRNA | BC003600.1 | - | 4, 5 | 627 | 3355972 | |
BC017673.1 | - | 4, 5 | 752 | 3893112 | ||
BC065818.1 | - | 4, 5 | 558 | 6196307 | ||
U24576.1 | - | 4, 5 | 1264 |