Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024616.1 | 1 | 431 | |||
GenBank mRNA | BC069671.1 | - | 1 | 354 | ||
BC069696.1 | - | 1 | 353 | |||
BC069772.1 | - | 1 | 353 | |||
BC112149.1 | - | 1 | 243 | |||
BC112175.1 | - | 1 | 243 | |||
Y10696.1 | - | 1 | 357 |