Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253908.1 | NP_001240837.1 | 9 | 989 | ||
NM_003739.5 | NP_003730.4 | 9 | 1012 | |||
GenBank mRNA | AB018580.1 | - | 9 | 990 | ||
AF149416.2 | - | 9 | 954 | |||
AK290365.1 | - | 9 | 972 | |||
AK296829.1 | - | 9 | 1056 | |||
AK315411.1 | - | 9 | 971 | |||
BC001479.2 | - | 9 | 999 | 2988160 | ||
BC019230.1 | - | 9 | 969 | 3682448 | ||
BT007286.1 | - | 9 | 943 | |||
D17793.1 | - | 9 | 994 | |||
DQ269985.1 | - | 10 | 943 | |||
S68288.1 | - | 9 | 953 |