Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304472.1 | NP_001291401.1 | 13 | 1731 | ||
NM_001304473.1 | NP_001291402.1 | 14 | 1757 | |||
NM_001304474.1 | NP_001291403.1 | 13 | 1702 | |||
NM_003741.3 | NP_003732.2 | 13 | 1731 | |||
NR_130747.1 | 11 | 1889 | ||||
XM_011513254.1 | XP_011511556.1 | 13 | 1584 | |||
XM_011513256.1 | XP_011511558.1 | 11 | 1740 | |||
XM_017007388.1 | XP_016862877.1 | 12 | 1665 | |||
XM_017007389.1 | XP_016862878.1 | 12 | 1665 | |||
XM_017007390.1 | XP_016862879.1 | 12 | 1665 | |||
XM_017007391.1 | XP_016862880.1 | 12 | 1665 | |||
XM_017007392.1 | XP_016862881.1 | 13 | 1584 | |||
XM_017007393.1 | XP_016862882.1 | 12 | 1665 | |||
XM_017007394.1 | XP_016862883.1 | 11 | 1453 | |||
GenBank mRNA | AB208947.1 | - | 8 | 2282 | ||
AF076612.1 | - | 10 | 1023 | |||
AF209928.1 | - | 13 | 1731 | |||
AF209930.1 | - | 14 | 1757 | |||
AF283325.1 | - | 13 | 1702 | |||
AK074147.1 | - | 11 | 1642 | |||
AK293456.1 | - | 13 | 1453 | |||
AK300213.1 | - | 5 | 1073 | |||
AK300476.1 | - | 2 | 160 | |||
AY358926.1 | - | 13 | 1621 | |||
BC112345.1 | - | 13 | 1516 | 40008417 |